5-Bromopyridine-2-sulphonyl chloride 95%

BD26231-1G BD26231-100MG
BLDPBD26231-1GEA 282 EUR
BLDPBD26231-1G BLDPBD26231-100MG
5-Bromopyridine-2-sulphonyl chloride 95%
5-Bromopyridine-2-sulphonyl chloride

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₅H₃BrClNO₂S
Storage Temperature: Freezer
MDL Number: MFCD06739112
CAS Number: 874959-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR