4-(Methoxycarbonyl)furan-2-boronic acid pinacol ester 98%

BD264339-100MG BD264339-250MG
BLDPBD264339-100MGEA 41 EUR
BLDPBD264339-100MG BLDPBD264339-250MG
4-(Methoxycarbonyl)furan-2-boronic acid pinacol ester 98%
4-(Methoxycarbonyl)furan-2-boronic acid pinacol ester

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₂H₁₇BO₅
MW: 252.0700 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12407248
CAS Number: 1111096-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR