Piperazin-1-yl(quinoxalin-6-yl)methanone hydrochloride 95%

BD264561-250MG BD264561-100MG
BLDPBD264561-250MGEA 80.9 EUR
BLDPBD264561-250MG BLDPBD264561-100MG
Piperazin-1-yl(quinoxalin-6-yl)methanone hydrochloride 95%
Piperazin-1-yl(quinoxalin-6-yl)methanone hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₃H₁₅ClN₄O
Storage Temperature: Ambient
MDL Number: MFCD13619874
CAS Number: 1204298-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR