N-Propargyltrifluoroacetamide 98%

BD265954-5G BD265954-1G
BLDPBD265954-5GEA 173 EUR
BLDPBD265954-5G BLDPBD265954-1G
N-Propargyltrifluoroacetamide 98%
N-Propargyltrifluoroacetamide

Storage condition: Sealed in dry, 2-8°C

Formula: C₅H₄F₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD14638370
CAS Number: 14719-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR