2-Amino-5-iodopyridine 97%

BD2682-5G BD2682-1000G
BLDPBD2682-5GEA 8.4 EUR
BLDPBD2682-5G BLDPBD2682-1000G
2-Amino-5-iodopyridine 97%
2-Amino-5-iodopyridine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₅IN₂
MW: 220.01 g/mol
Boiling Pt: 271
Storage Temperature: Ambient
MDL Number: MFCD00160312
CAS Number: 20511-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR