N-(tert-Butoxycarbonyl)propargylamine 98%

BD26998-10G BD26998-5G
BLDPBD26998-10GEA 33.6 EUR
BLDPBD26998-10G BLDPBD26998-5G
N-(tert-Butoxycarbonyl)propargylamine 98%
N-(tert-Butoxycarbonyl)propargylamine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₁₃NO₂
MW: 155.20 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07367245
CAS Number: 92136-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR