5-Amino-7-azaindole 97%

BD27109-1G BD27109-250MG
BLDPBD27109-1GEA 75.6 EUR
BLDPBD27109-1G BLDPBD27109-250MG
5-Amino-7-azaindole 97%
5-Amino-7-azaindole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₇N₃
Storage Temperature: Ambient
MDL Number: MFCD06659683
CAS Number: 100960-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR