(S)-tert-Butyl 2-carbamothioylpyrrolidine-1-carboxylate 95%

BD27116-5G BD27116-250MG
BLDPBD27116-5GEA 535 EUR
BLDPBD27116-5G BLDPBD27116-250MG
(S)-tert-Butyl 2-carbamothioylpyrrolidine-1-carboxylate 95%
(S)-tert-Butyl 2-carbamothioylpyrrolidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₀H₁₈N₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD06738835
CAS Number: 101410-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR