Methyl-4-fluoro-1H-indole-2-carboxylate 95%

BD272702-250MG BD272702-5G
BLDPBD272702-250MGEA 7.4 EUR
BLDPBD272702-250MG BLDPBD272702-5G
Methyl-4-fluoro-1H-indole-2-carboxylate 95%
Methyl-4-fluoro-1H-indole-2-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD06653401
CAS Number: 113162-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR