Methyl-5-bromo-2-hydroxynicotinate 98%

BD27291-1G BD27291-500G
BLDPBD27291-1GEA 10.5 EUR
BLDPBD27291-1G BLDPBD27291-500G
Methyl-5-bromo-2-hydroxynicotinate 98%
Methyl 5-bromo-2-hydroxynicotinate

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₆BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD03844843
CAS Number: 120034-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR