1-Methylimidazole-5-carbaldehyde 98%

BD2730-10G BD2730-25G
BLDPBD2730-10GEA 258 EUR
BLDPBD2730-10G BLDPBD2730-25G
1-Methylimidazole-5-carbaldehyde 98%
1-Methylimidazole-5-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₆N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00030439
CAS Number: 39021-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR