Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoate 95%
|
|
BD273736-100MG
BD273736-250MG
BLDPBD273736-100MGEA
45.2
EUR
BLDPBD273736-100MG
BLDPBD273736-250MG
Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoate 95%
Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthoate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₈H₂₁BO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD17015822 CAS Number: 643094-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...