tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 96%

BD27591-10G BD27591-1G
BLDPBD27591-10GEA 361 EUR
BLDPBD27591-10G BLDPBD27591-1G
tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate 96%
tert-Butyl-3,8-diazabicyclo[3.2.1]octane-8-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₁H₂₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD17016708
CAS Number: 149771-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR