2,2-Pentamethylene-1,3-dioxolane 97%
|
|
BD27803-1G
BD27803-5G
BLDPBD27803-1GEA
50.4
EUR
BLDPBD27803-1G
BLDPBD27803-5G
2,2-Pentamethylene-1,3-dioxolane 97%
2,2-Pentamethylene-1,3-dioxolane
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₁₄O₂ MW: 142.196 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005409 CAS Number: 177-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...