4-Nitropyridine-2-carbonitrile 98%

BD27910-250MG BD27910-1G
BLDPBD27910-250MGEA 16.8 EUR
BLDPBD27910-250MG BLDPBD27910-1G
4-Nitropyridine-2-carbonitrile 98%
4-Nitropyridine-2-carbonitrile

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₃N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00191627
CAS Number: 19235-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR