6-Methylpyridine-2,3-diamine hydrochloride 95% (contains H₂O)

BD279569-5G BD279569-10G
BLDPBD279569-5GEA 167 EUR
BLDPBD279569-5G BLDPBD279569-10G
6-Methylpyridine-2,3-diamine hydrochloride 95% (contains H₂O)
6-Methylpyridine-2,3-diamine hydrochloride

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₁₀ClN₃
Storage Temperature: Ambient
MDL Number: MFCD18384241
CAS Number: 1956367-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR