4-Chloro-N-methylpicolinamide 97%
|
|
BD28072-25G
BD28072-500G
BLDPBD28072-25GEA
31.5
EUR
BLDPBD28072-25G
BLDPBD28072-500G
4-Chloro-N-methylpicolinamide 97%
4-Chloro-N-methylpicolinamide
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₇H₇ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD02185921 CAS Number: 220000-87-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...