Ethyl 5-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 97%
|
|
BD281239-250MG
BD281239-100MG
BLDPBD281239-250MGEA
85.1
EUR
BLDPBD281239-250MG
BLDPBD281239-100MG
Ethyl 5-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate 97%
Ethyl 5-bromopyrazolo[1,5-a]pyrimidine-3-carboxylate
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₉H₈BrN₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD26518651 CAS Number: 1436686-17-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...