2-Amino-6-bromo-3-methylpyridine 97%

BD282012-1G BD282012-250MG
BLDPBD282012-1GEA 50.4 EUR
BLDPBD282012-1G BLDPBD282012-250MG
2-Amino-6-bromo-3-methylpyridine 97%
2-Amino-6-bromo-3-methylpyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₇BrN₂
MW: 187.037 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17014961
CAS Number: 89466-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR