(4-Chlorophenyl)(1H-pyrrol-2-yl)methanone 95%

BD283574-1G BD283574-250MG
BLDPBD283574-1GEA 451 EUR
BLDPBD283574-1G BLDPBD283574-250MG
(4-Chlorophenyl)(1H-pyrrol-2-yl)methanone 95%
(4-Chlorophenyl)(1H-pyrrol-2-yl)methanone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD00090258
CAS Number: 13169-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR