4-Chloro-3-quinolinamine 98%

BD284558-10G BD284558-25G
BLDPBD284558-10GEA 339 EUR
BLDPBD284558-10G BLDPBD284558-25G
4-Chloro-3-quinolinamine 98%
4-Chloro-3-quinolinamine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₉H₇ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD00234511
CAS Number: 58401-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR