2-Amino-5-iodo-4-methylpyridine 98%

BD28480-25G BD28480-10G
BLDPBD28480-25GEA 210 EUR
BLDPBD28480-25G BLDPBD28480-10G
2-Amino-5-iodo-4-methylpyridine 98%
2-Amino-5-iodo-4-methylpyridine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₆H₇IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD06637727
CAS Number: 356561-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR