tert-Butyl (5-bromopyridin-3-yl)carbamate 95%
|
|
BD28495-10G
BD28495-100G
BLDPBD28495-10GEA
118
EUR
BLDPBD28495-10G
BLDPBD28495-100G
tert-Butyl (5-bromopyridin-3-yl)carbamate 95%
tert-Butyl (5-bromopyridin-3-yl)carbamate
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₀H₁₃BrN₂O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD07367900 CAS Number: 361550-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...