2-Amino-5-thiazolecarboxylic acid 95%

BD28636-10G BD28636-25G
BLDPBD28636-10GEA 35.7 EUR
BLDPBD28636-10G BLDPBD28636-25G
2-Amino-5-thiazolecarboxylic acid 95%
2-Amino-5-thiazolecarboxylic acid

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₄H₄N₂O₂S
MW: 144.1500 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203554
CAS Number: 40283-46-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR