2,4-Dichloro-5-nitropyridine 98%
|
|
BD28761-100MG
BD28761-5G
BLDPBD28761-100MGEA
7.4
EUR
BLDPBD28761-100MG
BLDPBD28761-5G
2,4-Dichloro-5-nitropyridine 98%
2,4-Dichloro-5-nitropyridine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₅H₂Cl₂N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD07368834 CAS Number: 4487-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...