2-(2,2-Dimethylpropionylamino)isonicotinamide 95%

BD28794-250MG BD28794-1G
BLDPBD28794-250MGEA 107 EUR
BLDPBD28794-250MG BLDPBD28794-1G
2-(2,2-Dimethylpropionylamino)isonicotinamide 95%
2-(2,2-Dimethylpropionylamino)isonicotinamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₁H₁₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04039861
CAS Number: 470463-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR