4-Chloro-2-(methylthio)pyrimidine 98%

BD28853-5G BD28853-1G
BLDPBD28853-5GEA 9.5 EUR
BLDPBD28853-5G BLDPBD28853-1G
4-Chloro-2-(methylthio)pyrimidine 98%
4-Chloro-2-(methylthio)pyrimidine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₅ClN₂S
Storage Temperature: Refrigerator
MDL Number: MFCD00006083
CAS Number: 49844-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR