(R)-tert-Butyl 3-(methoxymethyl)piperazine-1-carboxylate 95%

BD290702-1G BD290702-250MG
BLDPBD290702-1GEA 1220 EUR
BLDPBD290702-1G BLDPBD290702-250MG
(R)-tert-Butyl 3-(methoxymethyl)piperazine-1-carboxylate 95%
(R)-tert-Butyl 3-(methoxymethyl)piperazine-1-carboxylate

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₁₁H₂₂N₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD19688482
CAS Number: 1359658-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR