(S)-Methyl 2-amino-3-cyclohexylpropanoate hydrochloride 97%
|
|
BD290708-5G
BD290708-100G
BLDPBD290708-5GEA
26.3
EUR
BLDPBD290708-5G
BLDPBD290708-100G
(S)-Methyl 2-amino-3-cyclohexylpropanoate hydrochloride 97%
(S)-Methyl 2-amino-3-cyclohexylpropanoate hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₀H₂₀ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00153486 CAS Number: 17193-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...