3-(2-Chloropyrimidin-4-yl)-1-methyl-1H-indole 98%
|
|
BD291427-25G
BD291427-10G
BLDPBD291427-25GEA
51.5
EUR
BLDPBD291427-25G
BLDPBD291427-10G
3-(2-Chloropyrimidin-4-yl)-1-methyl-1H-indole 98%
3-(2-Chloropyrimidin-4-yl)-1-methyl-1H-indole
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₃H₁₀ClN₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD26397861 CAS Number: 1032452-86-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...