tert-Butyl (3-hydroxycyclohexyl)carbamate 97%

BD29221-250MG BD29221-25G
BLDPBD29221-250MGEA 20 EUR
BLDPBD29221-250MG BLDPBD29221-25G
tert-Butyl (3-hydroxycyclohexyl)carbamate 97%
tert-Butyl (3-hydroxycyclohexyl)carbamate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₁H₂₁NO₃
Storage Temperature: Refrigerator
MDL Number: MFCD06658389
CAS Number: 610302-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR