(1R,2R)-trans-2-(Boc-amino)cyclohexanol hydrochloride 97%
|
|
BD292919-250MG
BD292919-10G
BLDPBD292919-250MGEA
18.9
EUR
BLDPBD292919-250MG
BLDPBD292919-10G
(1R,2R)-trans-2-(Boc-amino)cyclohexanol hydrochloride 97%
(1R,2R)-trans-2-(Boc-amino)cyclohexanol hydrochloride
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₁H₂₁NO₃ MW: 215.2893 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11656036 CAS Number: 155975-19-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...