(2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol 97%

BD294229-1G BD294229-250MG
BLDPBD294229-1GEA 22.1 EUR
BLDPBD294229-1G BLDPBD294229-250MG
(2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol 97%
(2R,3R)-2-(2,4-Difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butane-2,3-diol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₃F₂N₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28902278
CAS Number: 133775-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR