2-Acetyl-6-methylpyridine 98%
|
|
BD29436-1G
BD29436-5G
BLDPBD29436-1GEA
13.7
EUR
BLDPBD29436-1G
BLDPBD29436-5G
2-Acetyl-6-methylpyridine 98%
2-Acetyl-6-methylpyridine
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₈H₉NO Storage Temperature: Ambient |
MDL Number:
MFCD06658293 CAS Number: 6940-57-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...