2-(1-(Ethylsulphonyl)azetidin-3-ylidene)acetonitrile 97%

BD295152-1G BD295152-5G
BLDPBD295152-1GEA 8.4 EUR
BLDPBD295152-1G BLDPBD295152-5G
2-(1-(Ethylsulphonyl)azetidin-3-ylidene)acetonitrile 97%
2-(1-(Ethylsulphonyl)azetidin-3-ylidene)acetonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₁₀N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD22987586
CAS Number: 1187595-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR