4,6-Dichloropyridin-3-amine 97%

BD29518-250MG BD29518-5G
BLDPBD29518-250MGEA 25.2 EUR
BLDPBD29518-250MG BLDPBD29518-5G
4,6-Dichloropyridin-3-amine 97%
4,6-Dichloropyridin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₅H₄Cl₂N₂
Storage Temperature: Refrigerator
MDL Number: MFCD00234054
CAS Number: 7321-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR