tert-Butyl 3-(4-bromo-1H-pyrazol-1-yl)azetidine-1-carboxylate 98%
|
|
BD295184-1G
BD295184-250MG
BLDPBD295184-1GEA
207
EUR
BLDPBD295184-1G
BLDPBD295184-250MG
tert-Butyl 3-(4-bromo-1H-pyrazol-1-yl)azetidine-1-carboxylate 98%
tert-Butyl 3-(4-bromo-1H-pyrazol-1-yl)azetidine-1-carboxylate
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₁H₁₆BrN₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD17480368 CAS Number: 877399-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...