Methyl-4-amino-2-fluorobenzoate 98%

BD29532-500G BD29532-10G
BLDPBD29532-500GEA 1550 EUR
BLDPBD29532-500G BLDPBD29532-10G
Methyl-4-amino-2-fluorobenzoate 98%
Methyl 4-amino-2-fluorobenzoate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₈FNO₂
Storage Temperature: Ambient
MDL Number: MFCD09037917
CAS Number: 73792-08-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR