1-(3-Fluorophenyl)cyclopropanamine 98%

BD29582-5G BD29582-1G
BLDPBD29582-5GEA 451 EUR
BLDPBD29582-5G BLDPBD29582-1G
1-(3-Fluorophenyl)cyclopropanamine 98%
1-(3-Fluorophenyl)cyclopropanamine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₉H₁₀FN
Storage Temperature: Refrigerator
MDL Number: MFCD07374478
CAS Number: 764647-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR