4-Cyano-3-methoxybenzoic acid 95%
|
|
BD297207-250MG
BD297207-5G
BLDPBD297207-250MGEA
116
EUR
BLDPBD297207-250MG
BLDPBD297207-5G
4-Cyano-3-methoxybenzoic acid 95%
4-Cyano-3-methoxybenzoic acid
Storage condition: Sealed in dry, Room Temperature
Formula:
C₉H₇NO₃ Storage Temperature: Ambient |
MDL Number:
MFCD18203531 CAS Number: 102362-00-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...