(S)-6-Methoxychroman-3-carboxylic acid 98%
|
|
BD297453-250MG
BD297453-100MG
BLDPBD297453-250MGEA
806
EUR
BLDPBD297453-250MG
BLDPBD297453-100MG
(S)-6-Methoxychroman-3-carboxylic acid 98%
(S)-6-Methoxychroman-3-carboxylic acid
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₁H₁₂O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD10696117 CAS Number: 182570-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...