3-Butyn-1-amine hydrochloride 95%

BD29809-1G BD29809-100G
BLDPBD29809-1GEA 47.3 EUR
BLDPBD29809-1G BLDPBD29809-100G
3-Butyn-1-amine hydrochloride 95%
3-Butyn-1-amine hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₇N·HCl
MW: 105.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658391
CAS Number: 88211-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR