4-Bromooxindole 97%

BD29976-250MG BD29976-100G
BLDPBD29976-250MGEA 8.4 EUR
BLDPBD29976-250MG BLDPBD29976-100G
4-Bromooxindole 97%
4-Bromooxindole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₈H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD06659911
CAS Number: 99365-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR