D-tert-Leucine (H-D-Tle-OH) 97%
|
|
BD3015-1G
BD3015-500G
BLDPBD3015-1GEA
7.4
EUR
BLDPBD3015-1G
BLDPBD3015-500G
D-tert-Leucine (H-D-Tle-OH) 97%
D-tert-Leucine (H-D-Tle-OH)
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₆H₁₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00004265 CAS Number: 26782-71-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...