tert-Butyl ethyl oxalate 98%

BD301615-250MG BD301615-1G
BLDPBD301615-250MGEA 25.2 EUR
BLDPBD301615-250MG BLDPBD301615-1G
tert-Butyl ethyl oxalate 98%
tert-Butyl ethyl oxalate

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₁₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD11052953
CAS Number: 50624-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR