2-Bromo-3-iodotoluene 95%

BD302153-1G BD302153-100MG
BLDPBD302153-1GEA 185 EUR
BLDPBD302153-1G BLDPBD302153-100MG
2-Bromo-3-iodotoluene 95%
2-Bromo-3-iodotoluene

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₆BrI
Storage Temperature: Refrigerator
MDL Number: MFCD11977389
CAS Number: 888214-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR