3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole 95%

BD303297-50MG BD303297-100MG
BLDPBD303297-50MGEA 35.7 EUR
BLDPBD303297-50MG BLDPBD303297-100MG
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole 95%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-((2-(trimethylsilyl)ethoxy)methyl)-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₅H₂₉BN₂O₃Si
MW: 324.29 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26403372
CAS Number: 1146162-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR