6,7-Difluoroquinolin-3-amine 95%

BD304224-100MG BD304224-1G
BLDPBD304224-100MGEA 275 EUR
BLDPBD304224-100MG BLDPBD304224-1G
6,7-Difluoroquinolin-3-amine 95%
6,7-Difluoroquinolin-3-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₆F₂N₂
Storage Temperature: Ambient
MDL Number: MFCD20723386
CAS Number: 318684-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR