D-Lysine 98%

BD3043-5G BD3043-250MG
BLDPBD3043-5GEA 89.3 EUR
BLDPBD3043-5G BLDPBD3043-250MG
D-Lysine 98%
D-Lysine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₁₄N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00008234
CAS Number: 923-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR