Methyl 3-(cyanomethyl)-1H-indole-5-carboxylate 97+%

BD304577-250MG BD304577-100MG
BLDPBD304577-250MGEA 66.2 EUR
BLDPBD304577-250MG BLDPBD304577-100MG
Methyl 3-(cyanomethyl)-1H-indole-5-carboxylate 97+%
Methyl 3-(cyanomethyl)-1H-indole-5-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₂H₁₀N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD09954797
CAS Number: 113438-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR